missing translation for 'onlineSavingsMsg'
Learn More

Sars E2 Antibody (4A6C9), Novus Biologicals™

Product Code. 18505822 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
Unit Size:
25µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18505822 0.025 mL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18505822 Supplier Novus Biologicals Supplier No. NBP1288500.025ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sars E2 Monoclonal specifically detects Sars E2 in SARS-CoV samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sars E2
Applications Western Blot
Classification Monoclonal
Clone 4A6C9
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry-Paraffin
Formulation Subclonal supernatant with No Preservative
Host Species Mouse
Immunogen Purified recombinant fragment of SARS-E2 glycoprotein precursor expressed in E. Coli.
Purification Method Tissue culture supernatant
Quantity 0.025 mL
Primary or Secondary Primary
Test Specificity Specific for SARS-E2.
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.