missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SARS-CoV-2 Spike S1 Protein Antibody (CR3022) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
£411.00 - £694.00
Specifications
| Antigen | SARS-CoV-2 Spike S1 Protein |
|---|---|
| Clone | CR3022 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30033838
|
Novus Biologicals™
NBP3-26919-0.025mg |
0.025 mg |
£411.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30039624
|
Novus Biologicals™
NBP3-26919-0.1mg |
0.1 mg |
£694.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SARS-CoV-2 Spike S1 Protein Monoclonal antibody specifically detects SARS-CoV-2 Spike S1 Protein in SARS-CoV-2 samples. It is validated for ELISASpecifications
| SARS-CoV-2 Spike S1 Protein | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS pH 7.5 | |
| Recombinant SARS-CoV-2 Spike S1 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| CR3022 | |
| ELISA | |
| Unconjugated | |
| Human | |
| SARS-CoV-2 | |
| SARS-CoV-2 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title