missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SARS-CoV-2 Spike S1 Protein Antibody (CR3022) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26919-0.025mg
This item is not returnable.
View return policy
Description
SARS-CoV-2 Spike S1 Protein Monoclonal antibody specifically detects SARS-CoV-2 Spike S1 Protein in SARS-CoV-2 samples. It is validated for ELISA
Specifications
| SARS-CoV-2 Spike S1 Protein | |
| Monoclonal | |
| Unconjugated | |
| PBS pH 7.5 | |
| Human | |
| Protein A purified | |
| RUO | |
| SARS-CoV-2 | |
| Purified |
| ELISA | |
| CR3022 | |
| ELISA 1:5000-1:10000 | |
| SARS-CoV-2 | |
| Recombinant SARS-CoV-2 Spike S1 Protein | |
| 0.025 mg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction