missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Spike Human anti-Virus, Unconjugated, Clone: AM005415, Proteintech
Human Recombinant Antibody
£177.00 - £530.00
Specifications
| Antigen | SARS-CoV-2 Spike |
|---|---|
| Clone | AM005415 |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16850446
|
Proteintech
91347-PTG-20UG |
20 μg |
£177.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16840446
|
Proteintech
91347-PTG-100UG |
100 μg |
£530.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SARS-CoV-2 Spike antibody from Proteintech is a human recombinant antibody generated with virus of virus SARS-CoV-2 Spike. This antibody recognizes virus antigen. The SARS-CoV-2 Spike antibody has been validated for the following applications: ELISA analysis.
Specifications
| SARS-CoV-2 Spike | |
| ELISA | |
| Unconjugated | |
| Virus | |
| IgG1 |
| AM005415 | |
| Monoclonal | |
| Human | |
| 43740568 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title