Learn More
SARS-CoV-2 Spike Human anti-Virus, Unconjugated, Clone: AM005415, Proteintech
Human Recombinant Antibody
Brand: Proteintech 91347-PTG-20UG
Description
The SARS-CoV-2 Spike antibody from Proteintech is a human recombinant antibody generated with virus of virus SARS-CoV-2 Spike. This antibody recognizes virus antigen. The SARS-CoV-2 Spike antibody has been validated for the following applications: ELISA analysis.
Specifications
| SARS-CoV-2 Spike | |
| Monoclonal | |
| Unconjugated | |
| 20 μg | |
| 43740568 | |
| IgG1 |
| ELISA | |
| AM005415 | |
| Human | |
| Primary | |
| Virus |