missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAPS3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £364.00
Specifications
| Antigen | SAPS3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
SAPS3 Polyclonal antibody specifically detects SAPS3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| SAPS3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 55291 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| C11orf23SAPLa, chromosome 11 open reading frame 23, DKFZp781E17107, DKFZp781E2374, DKFZp781O2362, FLJ11058, FLJ43065, KIAA1558MGC125711, PP6R3MGC125712, protein phosphatase 6, regulatory subunit 3, SAPLSAP190, SAPS domain family member 3, SAPS3SAPS domain family, member 3, serine/threonine-protein phosphatase 6 regulatory subunit 3, Sporulation-induced transcript 4-associated protein SAPL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human PPP6R3 (NP_060782.2). MFWKFDLHSSSHIDTLLEREDVTLKELMDEEDVLQECKAQNRKLIEFLLKAECLEDLVSFIIEEPPQDMDEKIRYKYPNISCELLTSDVSQMNDRLGEDESLLMKLYSFLLNDSPLNPLLASFFSKVLSILISRKPEQIVDFLKKKHDFVDLIIKHIGTSAIMDLLLRLLTCIEPPQPRQDVLNWLNEEKIIQRLVEIVHPSQEEDRHSNASQSLCEIVRLSRDQMLQIQNSTEPDPLLATLEKQEIIEQLLSNIFHKEK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title