missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAPS3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93106-0.1ml
This item is not returnable.
View return policy
Description
SAPS3 Polyclonal antibody specifically detects SAPS3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SAPS3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| C11orf23SAPLa, chromosome 11 open reading frame 23, DKFZp781E17107, DKFZp781E2374, DKFZp781O2362, FLJ11058, FLJ43065, KIAA1558MGC125711, PP6R3MGC125712, protein phosphatase 6, regulatory subunit 3, SAPLSAP190, SAPS domain family member 3, SAPS3SAPS domain family, member 3, serine/threonine-protein phosphatase 6 regulatory subunit 3, Sporulation-induced transcript 4-associated protein SAPL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human PPP6R3 (NP_060782.2). MFWKFDLHSSSHIDTLLEREDVTLKELMDEEDVLQECKAQNRKLIEFLLKAECLEDLVSFIIEEPPQDMDEKIRYKYPNISCELLTSDVSQMNDRLGEDESLLMKLYSFLLNDSPLNPLLASFFSKVLSILISRKPEQIVDFLKKKHDFVDLIIKHIGTSAIMDLLLRLLTCIEPPQPRQDVLNWLNEEKIIQRLVEIVHPSQEEDRHSNASQSLCEIVRLSRDQMLQIQNSTEPDPLLATLEKQEIIEQLLSNIFHKEK | |
| 0.1 mL | |
| Signal Transduction | |
| 55291 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction