missing translation for 'onlineSavingsMsg'
Learn More

SAPS2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£149.00 - £366.00

Specifications

Antigen SAPS2
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18673790
View Documents
Novus Biologicals
NBP2-94580-0.02ml
0.02 mL
£149.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18699860
View Documents
Novus Biologicals
NBP2-94580-0.1ml
0.1 mL
£366.00
0.01mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SAPS2 Polyclonal antibody specifically detects SAPS2 in Human samples. It is validated for Western Blot
Specifications

Specifications

SAPS2
Western Blot
Unconjugated
Rabbit
Signal Transduction
PBS (pH 7.3), 50% glycerol
9701
IgG
Affinity purified
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
Human
dJ579N16.1, KIAA0685PP6R2SAPS2serine/threonine-protein phosphatase 6 regulatory subunit 2, protein phosphatase 6, regulatory subunit 2, SAP190, SAPS domain family member 2, SAPS domain family, member 2
Recombinant fusion protein containing a sequence corresponding to amino acids 640-959 of human PPP6R2 (NP_001229827.1). EDSDTRCAARVMARPRFGAPHASESCSKNGPERGGQDGKASLEAHRDAPGAGAPPAPGKKEAPPVEGDSEGAMWTAVFDEPANSTPTAPGVVRDVGSSVWAAGTSAPEEKGWAKFTDFQPFCCSESGPRCSSPVDTECSHAEGSRSQGPEKAFSPASPCAWNVCVTRKAPLLASDSSSSGGSHSEDGDQKAASAMDAVSRGPGREAPPLPTVARTEEAVGRVGCADSRLLSPACPAPKEVTAAPAVAVPPEATVAITTALSKAGPAIPTPAVSSALAVAVPLGPIMAVTAAPAMVATLGTVTKDGKTDAPPEGAALNGPV
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.