missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAPS2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94580-0.02ml
This item is not returnable.
View return policy
Description
SAPS2 Polyclonal antibody specifically detects SAPS2 in Human samples. It is validated for Western Blot
Specifications
| SAPS2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| dJ579N16.1, KIAA0685PP6R2SAPS2serine/threonine-protein phosphatase 6 regulatory subunit 2, protein phosphatase 6, regulatory subunit 2, SAP190, SAPS domain family member 2, SAPS domain family, member 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 640-959 of human PPP6R2 (NP_001229827.1). EDSDTRCAARVMARPRFGAPHASESCSKNGPERGGQDGKASLEAHRDAPGAGAPPAPGKKEAPPVEGDSEGAMWTAVFDEPANSTPTAPGVVRDVGSSVWAAGTSAPEEKGWAKFTDFQPFCCSESGPRCSSPVDTECSHAEGSRSQGPEKAFSPASPCAWNVCVTRKAPLLASDSSSSGGSHSEDGDQKAASAMDAVSRGPGREAPPLPTVARTEEAVGRVGCADSRLLSPACPAPKEVTAAPAVAVPPEATVAITTALSKAGPAIPTPAVSSALAVAVPLGPIMAVTAAPAMVATLGTVTKDGKTDAPPEGAALNGPV | |
| 0.02 mL | |
| Signal Transduction | |
| 9701 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction