missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAPS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57815
This item is not returnable.
View return policy
Description
SAPS1 Polyclonal specifically detects SAPS1 in Human samples. It is validated for Western Blot.
Specifications
| SAPS1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| PPP6R1 | |
| Synthetic peptides corresponding to PPP6R1 (protein phosphatase 6, regulatory subunit 1) Antibody(against the N terminal of PPP6R1. Peptide sequence MFWKFDLHTSSHLDTLLEREDLSLPELLDEEDVLQECKVVNRKLLDFLLQ. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Guinea pig: 100%; Equine: 100%; Human: 100%; Pig: 100%; Rabbit: 100%; Canine: 92%; Mouse: 92%; Rat: 92%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| KIAA1115MGC138185, PP6R1MGC142003, protein phosphatase 6, regulatory subunit 1, SAP190, SAPS domain family member 1, SAPS domain family, member 1, SAPS1serine/threonine-protein phosphatase 6 regulatory subunit 1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 22870 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction