missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ SAMSA Fluorescein, 5-((2-(and-3)-S-(acetylmercapto) succinoyl) amino) Fluorescein, mixed isomers

Product Code. 11534127
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11534127

Brand: Molecular Probes™ A685

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

SAMSA Fluorescein, 5-((2-(and-3)-S-(acetylmercapto) succinoyl) amino) Fluorescein, mixed isomers

SAMSA fluorescein is a useful reagent for forming fluorescent protein conjugates and for assaying maleimide and iodoacetamide moieties on proteins with fluorescein. SAMSA fluorescein is activated with base to remove the acetyl protecting group, thereby generating a thiol-containing fluorescein.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Product Type SAMSA Fluorescein
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Thiol
Shipping Condition Room Temperature
Reactive Moiety Thiol
Label or Dye FITC (Fluorescein)
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.