missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Sample Tube Set

Product Code. 10394291 Shop All Buchi Products
Change view
Click to view available options
Capacity (Metric):
300 mL
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10394291 300 mL Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10394291 Supplier BUCHI Supplier No. 037377
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Guarantee safety, precision and durability with these tubes. BUCHI Sample Tube Set is ideal for Kjeldahl and Hydrolysis Instruments.

  • Extended lifetime thanks to unique high-quality extra thick glass
  • Corrosion protection as a result of precise glass joint dimensions
  • Good sealing safeguards against hazardous fumes
  • Tube design eliminates hot spots and glass breakages
  • High accuracy with tight sealed joints
  • Reliability from automated sample processing
TRUSTED_SUSTAINABILITY

Specifications

Product Type Sample Tube Set
Capacity (Metric) 300 mL
For Use With (Application) Sample Volumes up to 200mL in Volume or 5g in Weight
For Use With (Equipment) Kjeldahl and Hydrolysis Instruments
Material Glass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.