Learn More
Thermo Scientific™ Sample Storage Starter Packs
Click to view available options
Includes:
0.5mL Tubes, Decapper, Reader
1.0mL Tubes, Decapper, Reader
Tubes and Decapper
Tubes, Decapper, HSX Reader
Tubes, Decapper, ST Reader
Unit Size:
1 set
Beskrivelse
Introduce automation into your workflow with Thermo Scientific™ sample storage starter packs. Choose from five different starter pack options: BASIC, ADVANCED, PREMIUM, PREMIUM 0.5 mL and PREMIUM 1.mL. All Starter Packs include Matrix™ ScrewTop Tri-coded Tubes with an 8-Channel Screw Cap Decapper. Advanced and Premium Starter Packs also include a barcode reader.
Engineered with the value of your samples in mind:
- Choose from five different starter pack options.
- Eliminate the need to reformat non-2D cryotube samples.
- Secure sample tracking with 2D, 1D, and human readable codes.
- Cap/decap a full rack of tubes in less than 1 minute.
- Scan 96 tubes in as little as 1 second with advanced and premium starter packs.
- Help ensure quality with ISO-7 class cleanroom– manufactured tubes with sterility assurance.
Tekniske data
Tekniske data
| Includes | Tubes, Decapper, ST Reader |
| Type | Advanced Starter Pack |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion