Learn More
Cole-Parmer™ Acid Washed Grinding Beads
Product Code. 12879045 Shop All SPEX Certiprep Products
Change view
Click to view available options
Description:
Molecular Biology Grade
No Grade
Unit Size:
200g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
12879045 No Grade 200g
12534166 Molecular Biology Grade 200g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12879045 Supplier Cole-Parmer™ Supplier No. 2160
Acid Washed Grinding Beads are treated to remove fine particles and contaminants.
  • Choose from silica or zirconia grinding beads to help remove fine particles and contaminant.
  • Using untreated beads which contain impurities can affect homogenization and biological processes.
  • Acid washing silica and zirconia grinding beads help remove fine particles and contaminant for cleaner samples.
  • Select silica beads for general grinding applications; zirconia beads have greater density and offer higher impact power.
  • General-purpose grinding beads are suitable for most basic cell lysis applications.
  • Choose RNase/DNase-free treated molecular biology-grade grinding beads to inactivate contaminating enzymes.
  • Low-binding grinding beads are coated to reduce nonspecific binding of nucleic acids and proteins.
Specifications
Product Type Grinding Beads
Description No Grade
For Use With (Application) Grinding
For Use With (Equipment) 2010 Geno/Grinder
Packaging Quantity 200 g
Particle Size 800 to 1000 μm