missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Acid Washed Grinding Beads
Acid Washed Grinding Beads are treated to remove fine particles and contaminants.
Brand: Cole-Parmer™ 2161
Code : 50
Additional Details : Weight : 0.80000kg
Description
- Choose from silica or zirconia grinding beads to help remove fine particles and contaminant.
- Using untreated beads which contain impurities can affect homogenization and biological processes.
- Acid washing silica and zirconia grinding beads help remove fine particles and contaminant for cleaner samples.
- Select silica beads for general grinding applications; zirconia beads have greater density and offer higher impact power.
- General-purpose grinding beads are suitable for most basic cell lysis applications.
- Choose RNase/DNase-free treated molecular biology-grade grinding beads to inactivate contaminating enzymes.
- Low-binding grinding beads are coated to reduce nonspecific binding of nucleic acids and proteins.
Specifications
Grinding Beads | |
Grinding | |
200 g |
Molecular Biology Grade | |
2010 Geno/Grinder | |
800 to 1000 μm |