missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SAMM50 Recombinant Rabbit Monoclonal Antibody (24GB1135)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£172.00 - £488.00

Specifications

Antigen SAMM50
Clone 24GB1135
Concentration 2 mg/mL
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30274975
GREENER_CHOICE
View Documents
Invitrogen™
MA557702
100 μL
£488.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275684
View Documents
Invitrogen™
MA557703
20 μL
£172.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SAMM50 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

SAMM50 is a component of the sorting and assembly machinery (SAM) complex of the outer mitochondrial membrane. The SAM complex has a role in integrating beta-barrel proteins into the outer mitochondrial membrane (Humphries et al, 2005).
Specifications

Specifications

SAMM50
2 mg/mL
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6AXV4, Q8BGH2, Q9Y512
25813, 300111, 68653
A synthesized peptide derived from human SAMM50 (aa 350-390).
Antibody
24GB1135
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
Samm50
1110030L07Rik; CGI-51; OMP85; SAM50; SAMM50; SAMM50 sorting and assembly machinery component; sorting and assembly machinery 50kDa; sorting and assembly machinery component 50 homolog; sorting and assembly machinery component 50 homolog (S. cerevisiae); sorting and assembly machinery component 50 homolog A; TOB55; Transformation-related gene 3 protein; TRG3; TRG-3; YNL026W
Samm50
Primary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.