missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAMM50 Recombinant Rabbit Monoclonal Antibody (24GB1135)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557703
This item is not returnable.
View return policy
Description
SAMM50 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
SAMM50 is a component of the sorting and assembly machinery (SAM) complex of the outer mitochondrial membrane. The SAM complex has a role in integrating beta-barrel proteins into the outer mitochondrial membrane (Humphries et al, 2005).
Specifications
| SAMM50 | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6AXV4, Q8BGH2, Q9Y512 | |
| Samm50 | |
| A synthesized peptide derived from human SAMM50 (aa 350-390). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1135 | |
| Unconjugated | |
| Samm50 | |
| 1110030L07Rik; CGI-51; OMP85; SAM50; SAMM50; SAMM50 sorting and assembly machinery component; sorting and assembly machinery 50kDa; sorting and assembly machinery component 50 homolog; sorting and assembly machinery component 50 homolog (S. cerevisiae); sorting and assembly machinery component 50 homolog A; TOB55; Transformation-related gene 3 protein; TRG3; TRG-3; YNL026W | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 25813, 300111, 68653 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction