missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Saline 0.45% Solution Tubes
Click to view available options
Quantity:
100/Pk.
Format:
Tube
Manufacturing Origin:
United States
Unit Size:
Pack of 100
Description
Save time when preparing your samples for automated identification and susceptibility systems using Thermo Scientific™ Saline 0.45% Solution Tubes.
Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
- Easily adjust inoculum density with a clear tube
- Enhance sample security while promoting safe handling with a reliable screw cap on each tube
- Achieve versatility and flexibility with fill volumes in 1.8mL and 3.0mL
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Color | Clear liquid colorless |
| Content And Storage | 2 to 25°C |
| Format | Tube |
| Manufacturing Origin | United States |
| Product Type | Buffer Solution |
| Fill Volume | 3.0 mL |
| Packaging Type | Plastic tube with screw cap |
| Quantity | 100/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction