missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SAIVI™ Rapid Antibody Labeling Kit, Alexa Fluor™ 750

Product Code. 10256363
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10256363 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10256363 Supplier Invitrogen™ Supplier No. S30046
  • ONLINE EXCLUSIVE PRICE
    £658.00
    £493.65 / Pack of 1
    Save £164.35
    24% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

SAIVI™ Rapid Antibody Labeling Kit, Alexa Fluor™ 750

The Alexa Fluor™ 750 SAIVI™ Antibody Labeling Kit provides a convenient means to label antibodies with an optimal degree of labeling for in vivo imaging applications (DOL; ∼2) over a 6-fold antibody concentration range with no adjustments in reaction volume, dye concentration, or antibody concentration necessary. Using this procedure, optimally labeled antibodies are ready for applications that require azide-free reagents, such as live-cell imaging or direct injection into animals.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Labeling Kit
Quantity 1 Kit
Content And Storage Store in refrigerator 2°C to 8°C and protect from light.
Detection Method Fluorescence
Excitation/Emission 749/775
Labeling Scale 0.5 to 3 mg
Shipping Condition Room Temperature
Product Line Alexa Fluor, SAIVI
Label or Dye Alexa Fluor 750
Color Infrared
Labeling Target Antibodies
Label Type Alexa Fluor Dyes
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.