missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Chemware™ PTFE Griffin Beakers

Product Code. 10165347
Change view
Click to view available options
Capacity (Metric):
1 mL
3 mL
5 mL
10 mL
30 mL
50 mL
100 mL
250 mL
400 mL
600 mL
1000 mL
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10165347 50 mL Each
15147826 1 mL Each
12346978 3 mL Each
12356978 5 mL Each
12366978 10 mL Each
12376978 30 mL Each
12396978 100 mL Each
12306988 250 mL Each
12316988 400 mL Each
12326988 600 mL Each
12336988 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 10165347 Supplier Saint Gobain Life Sciences Supplier No. D1069036

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemware™ PTFE Griffin Beakers are noncontaminating and inert to highly corrosive reagents such as hydrofluoric acid.

  • Nonwettable, unbreakable and heat resistant
  • Easy to clean
  • Broad spout for easy pouring
  • Can be heated on hotplate up to 288°C (550°F) when proper care is exercised
  • Specially molded bottom allows rapid heat transfer
  • Unlike most PTFE beakers, these will boil water when covered

Applications:

Ideal for digestion

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 1.69 oz.
Capacity (Metric) 50 mL
Material PTFE
For Use With (Application) Ideal for Digestion
Height (English) 2.05 in.
Height (Metric) 5.23 cm
Includes Spout
Temperature (English) Max. Operating 550°F
Temperature (Metric) Max. Operating 288°C
Type Beaker
Style Griffin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.