Learn More
Fisherbrand™ Safety Cover for Dry Block Heaters
Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
Fisherbrand™ 1 Block System
Fisherbrand™ 2 Block System
Fisherbrand™ 4 Block System
Unit Size:
Each
Description
- Made from tough clear acrylic for maximum visibility
- Prevents accidental touching of a hot block or contamination of samples from splashes
Specifications
Specifications
| For Use With (Equipment) | Fisherbrand™ 1 Block System |
| Type | Safety Cover |
| For Use With (Application) | Preventing Burns or Contamination |
| Material | Acrylic |
| Height (Metric) | 85 mm |