missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Safety Bottle Carrier

Product Code. 17810782 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Black
Blue
Green
Red
Size:
1.5 L
4.5 L
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Size Unit Size
17810782 Green 4.5 L Each
17880772 Green 1.5 L Each
17800782 Black 1.5 L Each
17890772 Blue 1.5 L Each
17820782 Red 1.5 L Each
17850782 Blue 4.5 L Each
17840782 Red 4.5 L Each
17830782 Black 4.5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 17810782
  • ONLINE EXCLUSIVE PRICE
    £112.00
    £74.04 / Pack of 1
    Save £37.96
    33% Off
View all promotions
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Safety Bottle Carrier provides a safe and reliable way to transport a variety of bottled reagents in your lab.

  • Robust Design
  • Ergonomic handle prevents hand fatigue and related accidents when carrying heavier loads
  • Handle can be pressed down to either side for easier loading and unloading
  • Centering ring stabilizes bottle in the center of carrier for safer, steadier transport
  • Carriers come in four colors with designated labeling areas for easy identification
  • Made of chemical resistant thermoplastic elastomer - PP blend

Specifications

Description Safety Bottle Carrier, 4.5 L, Green
Autoclavable Not Autoclavable
Color Green
Holds Bottles
Material Polypropylene
Positions 1
Shape Cylindrical
Stackable No
Sterility Non-sterile
For Use With (Application) Transportation of bottled regeants
For Use With (Equipment) 4.5 L bottles
Size 4.5 L
Temperature (Metric) Range -20 to 60°C
Length (Metric) 18.3 cm
Diameter (Metric) Opening 18.3 cm
Weight (Metric) .25 kg
Includes 1 Safety Bottle Carrier vessel and attachable handle
Product Type Bottle Holder
Diameter (Metric) Inner 18.3 cm
Shelves None
Height (Metric) 26.7 cm (40.6 cm Height, Handle Extended)
Array 1 x 1
Width (Metric) 18.3 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.