missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSF Progressive Safety™ Safety boots Quick Release 325mm Foundry Boot
Rubber nitrile sole
Brand: PSF Progressive Safety™ 117-8
Code : 91
Additional Details : Weight : 1.00000kg
Description
- 200 joule toe protection
- Manufactured to BS4676
- Rubber nitrile sole
- Sole resists oils acids, alkalis & heat to 300°C
- Black
- PSF Special Hazard
Specifications
8 |