missing translation for 'onlineSavingsMsg'
Learn More

PSF Progressive Safety™ Safety boots Quick Release 325mm Foundry Boot

Product Code. p-7182279
Click to view available options
Size:
5
6
7
8
9
10
12
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 12979393

Brand: PSF Progressive Safety™ 1175

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rubber nitrile sole

  • 200 joule toe protection
  • Manufactured to BS4676
  • Rubber nitrile sole
  • Sole resists oils acids, alkalis & heat to 300°C
  • Black
  • PSF Special Hazard

Specifications

Size 5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.