Learn More
Eppendorf™ SafeCode Plates (barcoded)
GREENER_CHOICE
Product Code. 17865938 Shop All Eppendorf Products
Change view
Click to view available options
Array:
16 x 24
Unit Size:
Pack of 80
Product Code. Array unitSize
17865938 16 x 24 Pack of 80
1 options
Product Code. 17865938 Supplier Eppendorf™ Supplier No. 0030113594
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Barcoded 96- or 384-well plates for reliable sample identification, efficient handling, and long-term storage.
  • 3-level coding ensures safe sample and vessel identification.
  • Codes on three sides of plates allow flexible reading.
  • ECC 200 error correction enables safe code reading even with up to 25% damaged codes.
  • Additionally, ID-specific information is available via Eppendorf DataPort for documentation.
  • Direct import into eLabInventory and eLabJournal™ software for convenient handling.
  • Import to other database systems is possible.
  • Manufactured from high-quality polypropylene for the highest sample purity.
TRUSTED_SUSTAINABILITY
Specifications
Barcode Includes Barcode
Format SBS Standard
Array 16 x 24
Product Line Eppendorf
Color White, Clear Bottom
Product Type PCR Plate
Material Polypropylene
Volume (Metric) Well 140 μL
No. of Wells 384
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Round, Round Bottom (U-well)
Quantity 80/Pk.
Fit for Purpose PCR Ready