missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Safe-Lock Tubes
GREENER_CHOICE

Product Code. 10430423 Shop All Eppendorf Products
Change view
Click to view available options
Capacity (Metric):
2 mL
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10430423 2 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10430423 Supplier Eppendorf™ Supplier No. 0030120230
Only null left
Add to basket
Edge
Add to basket

The hinged lid of Eppendorf Safe-Lock Tubes offers outstanding protection against unintentional opening, and completely eliminates any danger of leaks during incubation and storage.

  • Hinged Eppendorf Safe-Lock lid prevents unintentional lid opening during incubation and storage
  • Polypropylene provides high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results
  • g-Safe™: Depending on Safe-Lock Tube variant (see manual) exceptional centrifugation stability up to 30.000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
    - Precise lid sealing for minimal evaporation rates during long-term storage
  • Large frosted lid and surface on the side for easy labeling
  • Ensured functionality from -86 °C to 100 °C
  • Autoclavable when open (121 °C, 20 min)
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 2 mL
Color Dark Green
Packaging 1000
Temperature Range (Metric) -86 – 100°C

Eppendorf Safe-Lock Tubes are single use sample tubes intended as primary vessel in accordance with EN 14254:2004 and for storing human test specimen, with the exception of blood samples for in vitro diagnostics. Eppendorf Safe-Lock Tubes are intended for use by trained and skilled personnel only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.