missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | SAE1 |
|---|---|
| Concentration | 0.22 mg/mL |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16816483
|
Proteintech
10229-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16806483
|
Proteintech
10229-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Posttranslational modification of proteins by the addition of the small protein SUMO (see SUMO1), or sumoylation, regulates protein structure and intracellular localization. SAE1 and UBA2. form a heterodimer that functions as a SUMO-activating enzyme for the sumoylation of proteins (Okuma et al., 1999).Specifications
| SAE1 | |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q6AXQ0, Q9R1T2, Q9UBE0 | |
| 10055, 308384, 56459 | |
| SAE1 Fusion Protein Ag0275 | |
| Primary | |
| -20°C | |
| SAE1 |
| 0.22 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| AOS1, FLJ3091, HSPC140, SAE1, SUA1, UBLE1A | |
| Sae1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title