missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10229-1-AP-20UL
This item is not returnable.
View return policy
Description
Posttranslational modification of proteins by the addition of the small protein SUMO (see SUMO1), or sumoylation, regulates protein structure and intracellular localization. SAE1 and UBA2. form a heterodimer that functions as a SUMO-activating enzyme for the sumoylation of proteins (Okuma et al., 1999).Specifications
| SAE1 | |
| Polyclonal | |
| Unconjugated | |
| SAE1 | |
| AOS1, FLJ3091, HSPC140, SAE1, SUA1, UBLE1A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10055, 308384, 56459 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6AXQ0, Q9R1T2, Q9UBE0 | |
| Sae1 | |
| SAE1 Fusion Protein Ag0275 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction