missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sabouraud Glucose Chloramphenicol Selective Agar

Product Code. 10016732
This item is not returnable. View return policy
Product Code. 10016732 Supplier Thermo Scientific™ Supplier No. PO5070A
  • ONLINE EXCLUSIVE PRICE
    £10.30
    £10.06 / Pack of 10
    Save £0.24
    2% Off
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Isolate dermatophytes, other fungi and yeasts in clinical and environmental samples with this ready-to-use, selective medium.

Isolate dermatophytes, other fungi and yeasts in clinical and environmental samples with Thermo Scientific™ Oxoid™ Sabouraud Glucose Chloramphenicol Selective Agar. The acidic pH of the medium supports the growth of fungi while inhibiting bacterial growth in mixed samples. Fungi maintain their typical cultural appearance and thus may be readily identified according to the macroscopic characters described by Sabouraud1. The addition of chloramphenicol inhibits bacterial contaminants for the selective isolation of fungi.

Use Sabouraud Glucose Chloramphenicol Selective Agar for isolation of dermatophytes and other pathogenic fungi. The medium gives reliable results with Microsporum audouini, Microsporum canis, Trichophyton mentagrophytes, Trichophyton flavum, Trichophyton rubrum and Candida albicans2.

  • Suitable for identification of pathogenic fungi from material containing large numbers of other fungi or bacteria
  • Ready-to-use convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Color Ivory
Content And Storage 2°C to 10°C
Format 90mm Monoplate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Weight 17g ± 5%
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.