missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Sabouraud Dextrose Agar (EP/USP/JP/BP)
Click to view available options
Fill Weight:
100 to 101.5 g
250 to 254 g
450 to 454 g
Format:
125 mL Sirop Bottle
300 mL Sirop Bottle
500 mL Sirop Bottle
Unit Size:
Each
Pack of 10
Description
Isolate, cultivate, and enumerate fungi, particularly yeasts and molds, from clinical, food, cosmetic, and environmental samples using Sabouraud Dextrose Agar (EP/USP/JP/BP).
Specifications
Specifications
| Color | Straw |
| Content And Storage | 2°C to 25°C |
| Format | 500 mL Sirop Bottle |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar |
| Fill Weight | 450 to 454 g |
| Packaging Type | 10 Sirop Bottles with screw cap closures in a box |
| Quantity | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction