Learn More
Thermo Scientific™ Sabouraud Dextrose Agar (EP, USP, JP, BP)
Description
Isolate, cultivate and differentiate dermatophytes, other fungi and yeasts, using acidic pH Thermo Scientific™ Oxoid™ Sabouraud Dextrose Agar (EP, USP, JP, BP). The low pH supports the growth of fungi while inhibiting bacterial growth in mixed samples. Fungi maitain their typical appearance and thus may be readily identified according to standard macroscopic characters as described by Sabouraud1. The medium conforms to the formulations detailed in international pharmacopoeia2,3,4,5.
Use Sabouraud Dextrose Agar (EP, USP, JP, BP) for the isolation of dermatophytes, other fungi and yeasts. The medium gives reliable results with Microsporum audouini, Microsporum canis, Trichophyton mentagrophytes, Trichophyton flavum, Trichophyton rubrum and Candida albicans.
- Suitable for identification of pathogenic fungi from material containing large numbers of other fungi or bacteria
- Conforms to formulations detailed in International Pharmacopoeia2,3,4,5.
- Ready-to-use convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Color | Straw, clear |
| Content And Storage | 2°C to 10°C |
| Description | Sabouraud Dextrose Agar (EP, USP, JP, BP) |
| Format | 90mm Plate |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar |
| Fill Weight | 19.5g ± 1.0g |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.