missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sabouraud Dextrose Agar

Product Code. 10230103
Change view
Click to view available options
Quantity:
10/Pk.
Format:
90mm Monoplate
90mm Plate
Manufacturing Origin:
Germany
United Kingdom
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format Manufacturing Origin Unit Size
10230103 10/Pk. 90mm Monoplate Germany Pack of 10
11963662 10/Pk. 90mm Plate United Kingdom Pack of 10
12967158 10/Pk. 90mm Monoplate United Kingdom Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10230103 Supplier Thermo Scientific™ Supplier No. PO5001A
  • ONLINE EXCLUSIVE PRICE
    £10.00
    £9.06 / Pack of 10
    Save £0.94
    9% Off
View all promotions
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate, cultivate and differentiate dermatophytes, fungi and yeasts, such as Candida albicans, on a ready-to-use prepared media plate.

Isolate, cultivate and differentiate dermatophytes, fungi and yeasts, such as Candida albicans, using acidic pH Thermo Scientific™ Sabouraud Glucose Agar.

Fungi maintain their typical cultural appearance and may thus be readily identified according to the standard macroscopic characteristics. Infections caused by Candida spp. are diverse, ranging from superficial and local to deep-seated and disseminated. Candida albicans and Candida glabrata comprise 70-80% of yeast isolates recovered from patients with invasive disease. This culture media, also available in powder form, has applications in environmental monitoring and sterility testing, enabling corrective measures to be implemented rapidly if unacceptable levels of microbial loading indicate lapses in process control.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Tested in accordance with BP/EP/JP/USP.
Color Ivory, transparent
Content And Storage 2° to 12°C, in the dark
Format 90mm Monoplate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Weight 17 g ± 5 %
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.