missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- pH, Ion Concentration, and Redox/ORP benchtop meter that can display results in pH, mV, and Rel mV
- Durable and simple to operate with two buttons: 'Read' and 'Cal'
- IP54 Protection
- Comes with electrode arm that moves straight up and down, bringing the electrode into position where it works best for your sample
- Compatible with a wide range of electrodes for any application
- Compatible with ISM electrodes that capture and save calibration data, which is then automatically recognized by the meter once connected
- Meter connects to electrodes via BNC; S7 to BNC cables available
- Results can be captured by a printer with a time and date stamp
- Optional installation and qualification (IQ/OQ) package available through METTLER TOLEDO Service
Specifications
Specifications
| Accuracy (mV) | ±0.2 mV |
| Accuracy (pH) | ±0.002 |
| Description | S213 Kit: Micro Measurements |
| Channels | 1 |
| Display Type | TFT |
| Range (pH) | -2 to 20 |
| Resolution (pH) | 0.001/0.01/0.1 |
| Temperature Range (Metric) | -30.0°C to +130°C (MTC), -5.0°C to +130.0°C (ATC) |
| For Use With (Application) | Micro Measurements |
| Includes | S220 pH/Ion meter, InLab Ultra-Micro-ISM electrode, InLab 741-ISM electrode, Multipin-BNC/RCA electrode cable, electrode holder, 2 buffer sachets for pH 4.01, 7, 9.21 and 10.01, 2 standards sachets 1413 uS/cm, pH Measurement Guide, protective cover, test certificate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?