missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S1P5/EDG-8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£177.00 - £396.00
Specifications
| Antigen | S1P5/EDG-8 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18146631
|
Novus Biologicals
NBP2-24712SS |
0.025 mg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18789313
|
Novus Biologicals
NBP2-24712 |
0.1 mg |
£396.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
S1P5/EDG-8 Polyclonal specifically detects S1P5/EDG-8 in Human, Mouse, Rat, Canine, Primate, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| S1P5/EDG-8 | |
| Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Canine, Primate, Xenopus species | |
| Q9H228 | |
| 53637 | |
| A synthetic peptide corresponding to amino acids 15-55 of human Edg8 was used as the immunogen, GenBank no NP_110387. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| 0.2 ml PBS with 0.05% Sodium Azide | |
| Edg-8, EDG8S1P receptor Edg-8, Endothelial differentiation G-protein-coupled receptor 8, endothelial differentiation, sphingolipid G-protein-coupled receptor, 8, S1P receptor 5, S1P5, sphingosine 1-phosphate receptor 5, sphingosine 1-phosphate receptor EDG8, Sphingosine 1-phosphate receptor Edg-8, sphingosine-1-phosphate receptor 5, SPPR-1, SPPR-2 | |
| S1PR5 | |
| IgG | |
| Protein G purified | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a) and chimpanzee, 94% homologous in rhesus monkey, 84% homologous in cow and wild boar and 73% homologous in horse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title