missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S1P5/EDG-8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-24712
This item is not returnable.
View return policy
Description
S1P5/EDG-8 Polyclonal specifically detects S1P5/EDG-8 in Human, Mouse, Rat, Canine, Primate, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| S1P5/EDG-8 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS with 0.05% Sodium Azide | |
| Edg-8, EDG8S1P receptor Edg-8, Endothelial differentiation G-protein-coupled receptor 8, endothelial differentiation, sphingolipid G-protein-coupled receptor, 8, S1P receptor 5, S1P5, sphingosine 1-phosphate receptor 5, sphingosine 1-phosphate receptor EDG8, Sphingosine 1-phosphate receptor Edg-8, sphingosine-1-phosphate receptor 5, SPPR-1, SPPR-2 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a) and chimpanzee, 94% homologous in rhesus monkey, 84% homologous in cow and wild boar and 73% homologous in horse. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml | |
| Q9H228 | |
| S1PR5 | |
| A synthetic peptide corresponding to amino acids 15-55 of human Edg8 was used as the immunogen, GenBank no NP_110387. | |
| 0.1 mg | |
| GPCR | |
| 53637 | |
| Human, Mouse, Rat, Canine, Primate, Xenopus species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction