missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ S100P binding protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | S100P binding protein |
|---|---|
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066794
|
Novus Biologicals™
NBP3-30966-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066988
|
Novus Biologicals™
NBP3-30966-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
S100P binding protein Polyclonal antibody specifically detects S100P binding protein in Human samples. It is validated for ELISA,Immunohistochemistry,Western BlotSpecifications
| S100P binding protein | |
| Polyclonal | |
| Purified | |
| RUO | |
| Phosphate buffered saline | |
| 64766 | |
| IgG | |
| Antigen Affinity-purified |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DKFZp313K2325, FLJ12903, S100P binding protein, S100P-binding protein, S100P-binding protein Riken, S100PBPRS100P binding protein 1 | |
| Synthetic peptide derived from human S100P binding protein protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title