missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ S100P binding protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30966-100ug
This item is not returnable.
View return policy
Description
S100P binding protein Polyclonal antibody specifically detects S100P binding protein in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| S100P binding protein | |
| Polyclonal | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 64766 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| DKFZp313K2325, FLJ12903, S100P binding protein, S100P-binding protein, S100P-binding protein Riken, S100PBPRS100P binding protein 1 | |
| Synthetic peptide derived from human S100P binding protein protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction