missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S100A7A Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
£178.00 - £386.00
Specifications
| Antigen | S100A7A |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17763024
|
Invitrogen™
PA5121628 |
20 μL |
£178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17753024
![]() |
Invitrogen™
PA5121627 |
100 μL |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May be involved in epidermal differentiation and inflammation and might therefore be important for the pathogenesis of psoriasis and other diseases.Specifications
| S100A7A | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q86SG5 | |
| 338324 | |
| Ser2-Gln101 (Accession Q86SG5), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| S100A7A |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| NICE-2; Protein S100-A7A; S100 calcium binding protein A15; S100 calcium binding protein A7A; S100 calcium binding protein A7-like 1; S100 calcium-binding protein A15; S100 calcium-binding protein A7A; S100 calcium-binding protein A7-like 1; S100A15; S100A7A; S100A7f; S100A7L1 | |
| S100A7A | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title