missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S100A7A Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121627
This item is not returnable.
View return policy
Description
May be involved in epidermal differentiation and inflammation and might therefore be important for the pathogenesis of psoriasis and other diseases.
Specifications
| S100A7A | |
| Polyclonal | |
| Unconjugated | |
| S100A7A | |
| NICE-2; Protein S100-A7A; S100 calcium binding protein A15; S100 calcium binding protein A7A; S100 calcium binding protein A7-like 1; S100 calcium-binding protein A15; S100 calcium-binding protein A7A; S100 calcium-binding protein A7-like 1; S100A15; S100A7A; S100A7f; S100A7L1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 338324 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q86SG5 | |
| S100A7A | |
| Ser2-Gln101 (Accession Q86SG5), with N-terminal His Tag. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction