missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A6 Mouse, anti-Human, CoraLite Plus 488, Clone: 3D11C8, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | S100A6 |
|---|---|
| Clone | 3D11C8 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18060316
|
Proteintech
CL488-66098-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18000606
|
Proteintech
CL488-66098-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
The S100A6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human S100A6. This antibody recognizes human antigen. The S100A6 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Spezifikation
| S100A6 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 6277 | |
| IgG1 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 3D11C8 | |
| Immunofluorescence | |
| Liquid | |
| Human | |
| 2A9, 5B10, CABP, CACY, Calcyclin, MLN 4, PRA, Protein S100 A6, S100A6 | |
| Primary |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts