Learn More
S100A6 Mouse, anti-Human, CoraLite Plus 488, Clone: 3D11C8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66098-20UL
Description
The S100A6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human S100A6. This antibody recognizes human antigen. The S100A6 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| S100A6 | |
| 3D11C8 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| 2A9, 5B10, CABP, CACY, Calcyclin, MLN 4, PRA, Protein S100 A6, S100A6 | |
| 20 μL | |
| 6277 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |