missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A10 Mouse anti-Human, Clone: 1B3F9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66227-1-IG-150UL
This item is not returnable.
View return policy
Description
The S-100 family of calcium-activated proteins interact with a range of target proteins to modulate biological signaling pathways. Numerous cancer cell lines overexpress the plasminogen receptor S-100A10 on the extracellular cell surface, where it forms a heterotetrameric complex with Annexin II, though this association is not required for plasma membrane localization or binding and activation of plasminogen. Additionally, S-100A10 acts as a cellular chaperone for hepatitis B (Hep B) virus polymerase. Hep B virus polymerase normally localizes to the cytoplasm only, though in the presence of S-100A10 a portion relocates to the nucleus, implying a role for S-100A10 and intracellular calcium in the process of viral replication.Specifications
| S100A10 | |
| Monoclonal | |
| 1.72 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P60903 | |
| S100A10 | |
| S100A10 Fusion Protein Ag23596 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1B3F9 | |
| Unconjugated | |
| S100A10 | |
| 42C, ANX2L, ANX2LG, Ca[1], CAL1L, Calpactin 1 light chain, Calpactin I light chain, Cellular ligand of annexin II, CLP11, GP11, p10, p10 protein, P11, Protein S100 A10, S100A10 | |
| Mouse | |
| Protein G | |
| RUO | |
| 6281 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction