missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ S100A/B Antibody (SR3487), Novus Biologicals™
Rabbit Monoclonal Antibody
£240.00 - £290.00
Specifications
| Antigen | S100A/B |
|---|---|
| Clone | SR3487 |
| Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:50, Immunohistochemistry 1:100-1:500, Immunoprecipitation 1:50 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041661
|
Novus Biologicals™
NBP3-27774-50ul |
50 μL |
£240.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041600
|
Novus Biologicals™
NBP3-27774-100ul |
100 μL |
£290.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
S100A/B Monoclonal antibody specifically detects S100A/B in Human,Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western BlotSpecifications
| S100A/B | |
| Western Blot 1:500-1:1000, Flow Cytometry 1:50, Immunohistochemistry 1:100-1:500, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha | |
| A synthesized peptide derived from human S100A/B (Uniprot #: P23297) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR3487 | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 6271 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title