missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ S100A/B Antibody (SR3487), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27774-100ul
This item is not returnable.
View return policy
Description
S100A/B Monoclonal antibody specifically detects S100A/B in Human,Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| S100A/B | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| SR3487 | |
| Western Blot 1:500-1:1000, Flow Cytometry 1:50, Immunohistochemistry 1:100-1:500, Immunoprecipitation 1:50 | |
| protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha | |
| A synthesized peptide derived from human S100A/B (Uniprot #: P23297) | |
| 100 μL | |
| Breast Cancer, Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience | |
| 6271 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction