missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Elma™ Tec Clean S1 Cleaning Concentrate

Product Code. 15252768 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15252768 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15252768

Please to purchase this item. Need a web account? Register with us today!

Suitable for use in ultrasonic cleaning baths for nonferrous heavy metals, aluminum, stainless steel, plastics and glass. Fisherbrand™ Elma™ Tec Clean S1 Cleaning Concentrate is a mildly acidic cleaner.

  • Brass and copper alloys are brightened up
  • Easily removes oxide films, rust, lime and verdigris

Specifications

Form Liquid
pH 1.6
Composition Anionic surfactants <5%, non-ionic surfactants <5%, Benzisothiazolinone, Citric Acid
Concentration Cleaning 1 to 5% with water and for passivation 20 vol%
Container Type Can
Content And Storage -5°C to 25°C
Volume (Metric) 2.5 L
compliance-icons
Product Identifier
  • elma tec clean S1
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: alkylbenzenesulphonates, C10-13- alkylderivates, sodium salts, citric acid, monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.