missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (S)-SNAP 5114
GREENER_CHOICE

Product Code. 18035932
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18035932

Brand: Tocris Bioscience™ 1561/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

GABA uptake inhibitor

GABA transport inhibitor, showing selectivity for GAT-3 and GAT-2 (IC50 values are 5, 21 and 388 μM for hGAT-3, rGAT-2 and hGAT-1 respectively). Increases thalamic GABA levels and is an anticonvulsant following systemic administration in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 157604-55-2
Molecular Formula C30H35NO6

Specifications

Quantity 10 mg
Formula Weight 505.61
Purity >98%
Molecular Weight (g/mol) 505.61
Chemical Name or Material (S)-SNAP 5114
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.