accessibility menu, dialog, popup

UserName

CAS RN 157604-55-2

H3COOH3COCH3ONOHO

CAS RN 157604-55-2

IUPAC Name: 1-{2-[tris(4-methoxyphenyl)methoxy]ethyl}piperidine-3-carboxylic acid
Synonyms: 1-{2-[tris(4-methoxyphenyl)methoxy]ethyl}piperidine-3-carboxylic acid
Molecular Weight (g/mol): 505.61
Molecular Formula: C30H35NO6
InChi Key: VDLDUZLDZBVOAS-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1)C(OCCN1CCCC(C1)C(O)=O)(C1=CC=C(OC)C=C1)C1=CC=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results