missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress (S,R,S)-AHPC-PEG4-NH2 hydrochloride

Product Code. 16401019
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
10mg
1g
25mg
2g
500mg
50mg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16401019 1 g 1g
16494906 100 mg 100mg
16483868 10 mg 10mg
16460096 2 g 2g
16441566 500 mg 500mg
16412886 25 mg 25mg
16410247 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 16401019 Supplier MedChemExpress Supplier No. HY103604

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress (S,R,S)-AHPC-PEG4-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and 4-unit PEG linker used in PROTAC technology.

Specifications

Molecular Weight (g/mol) 700.29 g/mol
CAS 2064292-52-8
Content And Storage 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture)
Shipping Condition Room temperature
Quantity 1 g
Synonym VH032-PEG4-NH2 hydrochloride ∣ VHL Ligand-Linker Conjugates 4 hydrochloride ∣ E3 ligase Ligand-Linker Conjugates 7
Solubility H2O : 50 mg/mL (71.40 mM; Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.