missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress (S,R,S)-AHPC-PEG2-C4-Cl
Description
The connector of linker is Halogen group. (S,R,S)-AHPC-PEG2-C4-Cl incorporates the (S,R,S)-AHPC based VHL ligand and an alkyl/ether-based linker. (S,R,S)-AHPC-PEG2-C4-Cl is capable of inducing the degradation of GFP-HaloTag7 in cell-based assays.
Specifications
Specifications
| Molecular Weight (g/mol) | 651.26 g/mol |
| CAS | 1835705-57-1 |
| Content And Storage | Pure form -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Quantity | 2 g |
| Synonym | VH032-PEG2-C4-Cl ∣ VHL Ligand-Linker Conjugates 7 ∣ E3 ligase Ligand-Linker Conjugates 10 |
| Solubility | DMSO : ≥ 50 mg/mL (76.77 mM) ∣ Ethanol : 50 mg/mL (76.77 mM; Need ultrasonic) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?