missing translation for 'onlineSavingsMsg'
Learn More

S-Furfuryl thioacetate, 99%, Thermo Scientific™

Product Code. 11414857
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11414857

Brand: Thermo Scientific Alfa Aesar B21821.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13678-68-7
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.199
MDL Number MFCD00010083
InChI Key LQOUTUIIYXYBQW-UHFFFAOYSA-N
Synonym s-furfuryl thioacetate, furfuryl thioacetate, furfurylthiol acetate, ethanethioic acid, s-2-furanylmethyl ester, s-furfuryl ethanethioate, acetic acid, thio-, s-furfuryl ester, s-2-furanylmethyl ethanethioate, fema no. 3162, 2-furfurylthiol acetate, thioacetic acid s-furfuryl ester
PubChem CID 61660
IUPAC Name S-(furan-2-ylmethyl) ethanethioate
SMILES CC(=O)SCC1=CC=CO1

Specifications

Density 1.17
Boiling Point 211°C to 212°C
Flash Point 92°C (199°F)
Odor Stench
Refractive Index 1.525
Quantity 5 g
UN Number UN3334
Formula Weight 156.2
Percent Purity 99%
Chemical Name or Material S-Furfuryl thioacetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.